Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP2 Polyclonal Antibody

USP2 Polyclonal Antibody
Supplier: Thermo Scientific PA533100
Description
USP2 Polyclonal Antibody for IHC (P)

Specifications
USP2 | |
Polyclonal | |
Unconjugated | |
USP2 | |
41 kDa ubiquitin-specific protease; B930035K21Rik; deubiquitinating enzyme 2; MNCb-0190; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific peptidase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; ubiquitin thioesterase 2; ubiquitin thiolesterase 2; ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; Ubp41; Ubp69; UBP-t; USP2; USP9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 0.1% sodium azide | |
O75604 | |
USP2 | |
Synthetic 18 amino acid peptide from internal region of human USP2. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction