Learn More
USP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 103921AP150UL

Description
This antibody is specific to the isoform 1 (USP2a) of USP2.
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).Specifications
USP2 | |
Polyclonal | |
Unconjugated | |
USP2 | |
Deubiquitinating enzyme 2, ubiquitin specific peptidase 2, Ubiquitin thiolesterase 2, UBP41, USP2, USP9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115771, 53376, 9099 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75604, O88623, Q5U349 | |
Usp2 | |
USP2 Fusion Protein Ag0591 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.