Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP20 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059837AF594
Description
USP20 Polyclonal antibody specifically detects USP20 in Human, Mouse samples. It is validated for Western BlotSpecifications
USP20 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 20, EC 3.4.19.12, KIAA1003hVDU2, LSFR3A, pVHL-interacting deubiquitinating enzyme 2, ubiquitin carboxyl-terminal hydrolase 20, ubiquitin specific peptidase 20, ubiquitin specific protease 20, ubiquitin thioesterase 20, Ubiquitin thiolesterase 20, Ubiquitin-specific-processing protease 20, VDU2, VHL-interacting deubiquitinating enzyme 2 | |
The immunogen recognized by this antibody maps to a region between residue 863 and 913 of human ubiquitin specific peptidase 20 (pVHL-interacting deubiquitinating enzyme 2) using the numbering given in entry NP_006667.2 (GeneID 10868). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10868 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction