Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182946
Description
USP20 Polyclonal specifically detects USP20 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
USP20 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
Deubiquitinating enzyme 20, EC 3.4.19.12, KIAA1003hVDU2, LSFR3A, pVHL-interacting deubiquitinating enzyme 2, ubiquitin carboxyl-terminal hydrolase 20, ubiquitin specific peptidase 20, ubiquitin specific protease 20, ubiquitin thioesterase 20, Ubiquitin thiolesterase 20, Ubiquitin-specific-processing protease 20, VDU2, VHL-interacting deubiquitinating enzyme 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
USP20 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SDTDEKREGDRSPSEDEFLSCDSSSDRGEGDGQGRGGGSSQAETELLIPDEAGRAISEKERMKDRKFSWGQQRTNSEQVDEDADVDTAMAALDQPAEAQPPSPRSSSPCRTPEPDNDAHLRSSS | |
0.1 mL | |
GPCR | |
10868 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction