Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533102
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533102 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533102 Supplier Invitrogen™ Supplier No. PA533102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%); Orangutan, Gibbon, Rabbit (94%); Hamster (89%); Mouse, Bovine, Bat, Elephant, Panda, Horse, Pig, Opossum, Turkey, Chicken (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene USP20
Gene Accession No. Q9Y2K6
Gene Alias 1700055M05Rik; AI467231; Deubiquitinating enzyme 20; hVDU2; KIAA1003; LSFR3A; pVHL-interacting deubiquitinating enzyme 2; ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific peptidase 20; ubiquitin specific protease 20; ubiquitin thioesterase 20; ubiquitin thiolesterase 20; ubiquitin-specific-processing protease 20; Usp20; Vdu2; VHL-interacting deubiquitinating enzyme 2
Gene Symbols USP20
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human USP20.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10868
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.