Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582241
Description
Immunogen sequence: SDTDEKREGD RSPSEDEFLS CDSSSDRGEG DGQGRGGGSS QAETELLIPD EAGRAISEKE RMKDRKFSWG QQRTNSEQVD EDADVDTAMA ALDQPAEAQP PSPRSSSPCR TPEPDNDAHL RSSS.
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
USP20 | |
Polyclonal | |
Unconjugated | |
USP20 | |
1700055M05Rik; AI467231; Deubiquitinating enzyme 20; hVDU2; KIAA1003; LSFR3A; pVHL-interacting deubiquitinating enzyme 2; ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific peptidase 20; ubiquitin specific protease 20; ubiquitin thioesterase 20; ubiquitin thiolesterase 20; ubiquitin-specific-processing protease 20; Usp20; Vdu2; VHL-interacting deubiquitinating enzyme 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10868 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2K6 | |
USP20 | |
Recombinant protein corresponding to Human USP20. Recombinant protein control fragment (Product #RP-89266). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction