Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585719 Supplier Invitrogen™ Supplier No. PA585719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (89%), Rat (87%), Bovine (90%). Positive Control: 293T, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP20
Applications Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene USP20
Gene Accession No. Q9Y2K6
Gene Alias 1700055M05Rik; AI467231; Deubiquitinating enzyme 20; hVDU2; KIAA1003; LSFR3A; pVHL-interacting deubiquitinating enzyme 2; ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific peptidase 20; ubiquitin specific protease 20; ubiquitin thioesterase 20; ubiquitin thiolesterase 20; ubiquitin-specific-processing protease 20; Usp20; Vdu2; VHL-interacting deubiquitinating enzyme 2
Gene Symbols USP20
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP20. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.