Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP21 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | USP21 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
USP21 Polyclonal specifically detects USP21 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
USP21 | |
Unconjugated | |
RUO | |
Human | |
Deubiquitinating enzyme 21, EC 3.1.2.15, EC 3.4.19.12, MGC3394, ubiquitin carboxyl-terminal hydrolase 21, ubiquitin specific peptidase 21, ubiquitin specific protease 21, ubiquitin thioesterase 21, Ubiquitin thiolesterase 21, ubiquitin-specific processing protease 21, ubiquitin-specific protease 16, Ubiquitin-specific-processing protease 21, USP16, USP23NEDD8-specific protease | |
USP21 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Polyclonal | |
Rabbit | |
Cell Biology, Neuroscience, Ubiquitin Proteasome Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
27005 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CEAVNPTRFRAVFQKYVPSFSGYSQQDAQEFLKLLMERLHLEINRRGRRAPPILANGPVPSPPRRGGALLEEPELSDDDRANLMWKRYLEREDSKIVDLF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title