Learn More
Invitrogen™ USP21 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | USP21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | USP21 |
| Gene Accession No. | Q9UK80 |
| Gene Alias | deubiquitinating enzyme 21; ESTM28; NEDD8-specific protease; PP1490; similar to Ubiquitin carboxyl-terminal hydrolase 21 (Ubiquitin thiolesterase 21) (Ubiquitin-specific processing protease 21) (Deubiquitinating enzyme 21) (NEDD8-specific protease); ubiquitin carboxyl-terminal hydrolase 21; ubiquitin specific peptidase 21; ubiquitin specific protease 21; ubiquitin specific protease 23; ubiquitin thioesterase 21; ubiquitin thiolesterase 21; ubiquitin-specific processing protease 21; ubiquitin-specific protease 16; ubiquitin-specific protease 21; ubiquitin-specific-processing protease 21; USP16; USP21; Usp23; W53272 |
| Gene Symbols | USP21 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.