Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110556
Description
Immunogen sequence: MPQASEHRLG RTREPPVN.
This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described.
Specifications
| USP21 | |
| Polyclonal | |
| Unconjugated | |
| USP21 | |
| deubiquitinating enzyme 21; ESTM28; NEDD8-specific protease; PP1490; similar to Ubiquitin carboxyl-terminal hydrolase 21 (Ubiquitin thiolesterase 21) (Ubiquitin-specific processing protease 21) (Deubiquitinating enzyme 21) (NEDD8-specific protease); ubiquitin carboxyl-terminal hydrolase 21; ubiquitin specific peptidase 21; ubiquitin specific protease 21; ubiquitin specific protease 23; ubiquitin thioesterase 21; ubiquitin thiolesterase 21; ubiquitin-specific processing protease 21; ubiquitin-specific protease 16; ubiquitin-specific protease 21; ubiquitin-specific-processing protease 21; USP16; USP21; Usp23; W53272 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27005, 30941, 688466 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| B2GUX4, Q9QZL6, Q9UK80 | |
| USP21 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human USP21 (NP_0366073). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction