Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP22 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149644AF532
Description
USP22 Polyclonal specifically detects USP22 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
USP22 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L | |
Rabbit | |
0.1 mL | |
23326 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
USP22 | |
A genomic peptide made to an internal region of the human USP22 protein (within residues 100-205). [Swiss-Prot Q9H3R0] | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction