Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220829
Description
USP22 Polyclonal specifically detects USP22 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
USP22 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L | |
Rabbit | |
60 kDa | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers | |
23326 | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
Q9UPT9 | |
USP22 | |
Recombinant protein encompassing a sequence within the center region of human USP22. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (98%), Rat (98%), Bovine (99%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction