Learn More
Invitrogen™ USP22 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and Xenopus and zebrafish based on sequence homology.
Specifications
Specifications
| Antigen | USP22 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | USP22 |
| Gene Accession No. | Q9UPT9 |
| Gene Alias | AI427806; Deubiquitinating enzyme 22; KIAA1063; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific peptidase 22; ubiquitin specific protease 22; ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; ubiquitin-specific-processing protease 22; Usp22; USP3L |
| Gene Symbols | USP22 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.