Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP22 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988681
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8681 Supplier Bioss Supplier No. BS8611R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.

Specifications

Antigen USP22
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene USP22
Gene Accession No. Q9UPT9
Gene Alias AI427806; Deubiquitinating enzyme 22; Kiaa1063; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific peptidase 22; ubiquitin specific protease 22; Ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; Ubiquitin-specific-processing protease 22; Usp22; USP3L
Gene Symbols USP22
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human USP22.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.