Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | USP24 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
USP24 Polyclonal specifically detects USP24 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| USP24 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 23358 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RDSIPSEVDYETRQGVYSICLQLARFLLVGQTMPTLLDEDLTKDGIEALSSRPFRNVSRQTSRQMSLCGTPEKSSYRQLSVSDRSSIRVEEIIPAARVAIQTMEVS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 | |
| USP24 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title