Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP24 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10040830PE
Description
USP24 Polyclonal antibody specifically detects USP24 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
USP24 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 | |
Maps to a region between residue 2410 and the C-terminus (residue 2460) of human Ubiquitin Specific Peptidase 24 using the numbering given in entry XP_371254.5 (GeneID 23358). | |
0.1 mL | |
Cell Biology | |
23358 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction