Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP24 Antibody (SR1479), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32242850UL
Description
USP24 Monoclonal antibody specifically detects USP24 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
USP24 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1479 | |
Western Blot 1:500-1:2000 | |
Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 | |
A synthesized peptide derived from human USP24 (Uniprot #: Q9UPU5) | |
50 μg | |
Cell Biology | |
23358 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction