Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588001 Supplier Thermo Scientific Supplier No. PA588001
Add to Cart
Add to Cart

USP24 Polyclonal Antibody

USP24 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen USP24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene USP24
Gene Accession No. B1AY13, Q9UPU5
Gene Alias 2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; deubiquitinating enzyme 24; KIAA1057; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific peptidase 24; ubiquitin specific protease 24; ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; ubiquitin-specific-processing protease 24; Usp24
Gene Symbols USP24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2431-2620 of human USP24 (NP_0561212).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23358, 329908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.