Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP24 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1040830A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1040830A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1040830A30 Supplier Novus Biologicals Supplier No. NB10040830AF350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP24 Polyclonal specifically detects USP24 in Human samples. It is validated for Western Blot.

Specifications

Antigen USP24
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24
Gene Symbols USP24
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2410 and the C-terminus (residue 2460) of human Ubiquitin Specific Peptidase 24 using the numbering given in entry XP_371254.5 (GeneID 23358).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23358.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.