Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP24 Recombinant Rabbit Monoclonal Antibody (11E3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550040
Description
USP24 Recombinant Monoclonal Antibody for ICC/IF, Flow, ELISA
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP24 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
Specifications
USP24 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UPU5 | |
USP24 | |
A synthesized peptide derived from human USP24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry | |
11E3 | |
Unconjugated | |
USP24 | |
2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; deubiquitinating enzyme 24; KIAA1057; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific peptidase 24; ubiquitin specific protease 24; ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; ubiquitin-specific-processing protease 24; Usp24 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23358 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction