Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP24 Recombinant Rabbit Monoclonal Antibody (7K7N2)
GREENER_CHOICE

Catalog No. PIMA542612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542612 Supplier Invitrogen™ Supplier No. MA542612
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, Jurkat, SH-SY5Y, Mouse liver, Mouse thymus, Rat liver. Immunogen sequence: VKFLVTLAQK CPAAKEYFKE NSHHWSWAVQ WLQKKMSEHY WTPQSNVSNE TSTGKTFQRT ISAQDTLAYA TALLNEKEQS GSSNGSESSP ANENGDRHLQ Q.

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP24 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP24
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7K7N2
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene USP24
Gene Accession No. B1AY13, Q9UPU5
Gene Alias 2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; deubiquitinating enzyme 24; KIAA1057; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific peptidase 24; ubiquitin specific protease 24; ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; ubiquitin-specific-processing protease 24; Usp24
Gene Symbols USP24
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2500-2600 of human USP24 (Q9UPU5).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23358, 313427, 329908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.