Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP26 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | USP26 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
USP26 Polyclonal specifically detects USP26 in Human samples. It is validated for Western Blot.Specifications
| USP26 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Deubiquitinating enzyme 26, EC 3.4.19.12, MGC120066, MGC120067, MGC120068, ubiquitin carboxyl-terminal hydrolase 26, ubiquitin specific peptidase 26, ubiquitin specific protease 26, ubiquitin thioesterase 26, Ubiquitin thiolesterase 26, ubiquitin-specific processing protease 26, Ubiquitin-specific-processing protease 26 | |
| USP26 | |
| IgG | |
| 104 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9BXU7 | |
| 83844 | |
| Synthetic peptides corresponding to USP26 (ubiquitin specific peptidase 26) The peptide sequence was selected from the middle region of USP26 (NP_114113). Peptide sequence DILDKIKSKAKETKRNDDKGDHTYRLISVVSHLGKTLKSGHYICDAYDFE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title