Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145201
Description
Immunogen sequence: VPENPKRKKY VKTSKFVAFD RIINPTKDLY EDKNIRIPER FQKVSEQTQQ CDGMRICEQA PQQALPQSFP KPGTQGHTKN LLRP Antigen sequence identity: Mouse 30%; Rat 30%.
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases and is a deubiquitinating enzyme (DUB) with His and Cys domains. It is specifically expressed in testis tissue. Mutations in this gene have been associated with Sertoli cell-only syndrome and male infertility.
Specifications
USP26 | |
Polyclonal | |
Unconjugated | |
USP26 | |
Deubiquitinating enzyme 26; RGD1563443; Ubiquitin carboxyl-terminal hydrolase 26; ubiquitin specific peptidase 26; ubiquitin specific protease 26; ubiquitin thioesterase 26; ubiquitin thiolesterase 26; ubiquitin-specific processing protease 26; ubiquitin-specific-processing protease 26; USP26 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83844 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXU7 | |
USP26 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 692-775. Recombinant protein control fragment (Product #RP-110233). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction