Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531104 Supplier Invitrogen™ Supplier No. PA531104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases and is a deubiquitinating enzyme (DUB) with His and Cys domains. It is specifically expressed in testis tissue. Mutations in this gene have been associated with Sertoli cell-only syndrome and male infertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP26
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene USP26
Gene Accession No. Q9BXU7
Gene Alias Deubiquitinating enzyme 26; RGD1563443; Ubiquitin carboxyl-terminal hydrolase 26; ubiquitin specific peptidase 26; ubiquitin specific protease 26; ubiquitin thioesterase 26; ubiquitin thiolesterase 26; ubiquitin-specific processing protease 26; ubiquitin-specific-processing protease 26; USP26
Gene Symbols USP26
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human USP26. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.