Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP27X Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571973
Description
USP27X Polyclonal Antibody for Western Blot, ICC/IF
Deubiquitinase that can reduce the levels of BCL2L11/BIM ubiquitination and stabilize BCL2L11 in response to the RAF-MAPK-degradation signal. By acting on BCL2L11 levels, may counteract the anti-apoptotic effects of MAPK activity.
Specifications
USP27X | |
Polyclonal | |
Unconjugated | |
USP27X | |
Deubiquitinating enzyme 27; RGD1564628; Sfc11; Ubiquitin carboxyl-terminal hydrolase 22-like; Ubiquitin carboxyl-terminal hydrolase 27; ubiquitin carboxyl-terminal hydrolase 51-like; ubiquitin specific peptidase 27 X-linked; ubiquitin specific peptidase 27, X chromosome; ubiquitin specific peptidase 27, X-linked; ubiquitin specific peptidase 27, X-linked, pseudogene 1; ubiquitin specific protease 27, X chromosome; ubiquitin thioesterase 27; ubiquitin thiolesterase 22; ubiquitin thiolesterase 27; ubiquitin-specific-processing protease 27; USP22L; USP27; Usp27x; Usp27x-ps1; X-linked ubiquitin carboxyl-terminal hydrolase 27; X-linked ubiquitin thioesterase 27 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
389856 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
A6NNY8 | |
USP27X | |
KLH conjugated synthetic peptide between 1-30 amino acids of human USP27X. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction