Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP27X Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567449 Supplier Invitrogen™ Supplier No. PA567449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFYHKQVLEHE SEKVKEMNTQ AY Highest antigen sequence identity to the following orthologs - mouse 83%, rat 91%.

Deubiquitinase that can reduce the levels of BCL2L11/BIM ubiquitination and stabilize BCL2L11 in response to the RAF-MAPK-degradation signal. By acting on BCL2L11 levels, may counteract the anti-apoptotic effects of MAPK activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP27X
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP27X
Gene Accession No. A6NNY8
Gene Alias Deubiquitinating enzyme 27; RGD1564628; Sfc11; Ubiquitin carboxyl-terminal hydrolase 22-like; Ubiquitin carboxyl-terminal hydrolase 27; ubiquitin carboxyl-terminal hydrolase 51-like; ubiquitin specific peptidase 27 X-linked; ubiquitin specific peptidase 27, X chromosome; ubiquitin specific peptidase 27, X-linked; ubiquitin specific peptidase 27, X-linked, pseudogene 1; ubiquitin specific protease 27, X chromosome; ubiquitin thioesterase 27; ubiquitin thiolesterase 22; ubiquitin thiolesterase 27; ubiquitin-specific-processing protease 27; USP22L; USP27; Usp27x; Usp27x-ps1; X-linked ubiquitin carboxyl-terminal hydrolase 27; X-linked ubiquitin thioesterase 27
Gene Symbols USP27X
Host Species Rabbit
Immunogen Recombinant Human USP27X. Recombinant protein control fragment (Product #RP-108110).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.