Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP27X Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2837450.1ML
Description
USP27X Polyclonal specifically detects USP27X in Mouse samples. It is validated for Western Blot.Specifications
| USP27X | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 389856 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Deubiquitinating Enzyme 27, MRX105, Ubiquitin Carboxyl-Terminal Hydrolase 22-Like, Ubiquitin Carboxyl-Terminal Hydrolase 27, Ubiquitin Specific Peptidase 27, X-Linked, Ubiquitin Specific Protease 27, X Chromosome, Ubiquitin Specific Protease 27, X-Linked, Ubiquitin Thioesterase 27, Ubiquitin-Specific-Processing Protease 27, USP22L, USP27, X-Linked Ubiquitin Carboxyl-Terminal Hydrolase 27 | |
| The immunogen is a synthetic peptide directed towards the middle region of mouse USP27X | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction