Learn More
Invitrogen™ USP28 Monoclonal Antibody (A1F9)

Description
USP28 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | USP28 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | A1F9 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | USP28 |
| Gene Accession No. | Q5I043, Q96RU2 |
| Gene Alias | 9830148O20Rik; AU022237; Deubiquitinating enzyme 28; Kiaa1515; mKIAA1515; ubiquitin carboxyl-terminal hydrolase 28; ubiquitin carboxyl-terminal hydrolase 28 variant 1; ubiquitin specific peptidase 28; ubiquitin specific protease 28; ubiquitin thioesterase 28; ubiquitin thiolesterase 28; ubiquitin-specific-processing protease 28; Usp28 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.