Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140285 Supplier Invitrogen™ Supplier No. PA5140285
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SQGMAEQLQQ CIEESIIDEF LQQAPPPGVR KLDAQEHTEE TLNQSTELRL QKADLNHLGA LGSDNPGNKN ILDAENTRGE AKELTRNVKM G Highest antigen sequence indentity to the following orthologs: Rat - 40%, Mouse - 38%.

USP29 (Ubiquitin Specific Peptidase 29) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include cysteine-type endopeptidase activity and thiol-dependent ubiquitin-specific protease activity. An important paralog of this gene is USP37. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP29
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP29
Gene Accession No. Q9HBJ7
Gene Alias ASL1/Usp29 fusion; deubiquitinating enzyme 29; HOM-TES-84/86; Ocat; ossification center-associated transcript; ubiquitin carboxyl-terminal hydrolase 29; ubiquitin specific peptidase 29; ubiquitin specific protease 29; ubiquitin thioesterase 29; ubiquitin thiolesterase 29; ubiquitin-specific processing protease; ubiquitin-specific-processing protease 29; Usp29
Gene Symbols USP29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP29. Recombinant protein control fragment (Product #RP-109559).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.