Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077284UV
Description
USP3 Polyclonal antibody specifically detects USP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
USP3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 3, EC 3.1.2.15, EC 3.4.19.12, MGC129878, MGC129879, SIH003, ubiquitin carboxyl-terminal hydrolase 3, ubiquitin specific peptidase 3, ubiquitin specific protease 3, ubiquitin thioesterase 3, Ubiquitin thiolesterase 3, Ubiquitin-specific-processing protease 3, UBP | |
The immunogen recognized by this antibody maps to a region between residue 471 and 521 of human ubiquitin specific protease 3 using the numbering given in entry AAD42992.1 (GeneID 9960). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9960 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction