Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585512 Supplier Invitrogen™ Supplier No. PA585512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (96%), Xenopus laevis (87%), Rhesus Monkey (99%). Positive Control: 293T, A431, HeLa, human USP3-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene USP3
Gene Accession No. Q9Y6I4
Gene Alias AA409661; BC017156; Deubiquitinating enzyme 3; SIH003; Ubiquitin carboxyl-terminal hydrolase 3; ubiquitin specific peptidase 3; ubiquitin specific protease 3; ubiquitin thioesterase 3; ubiquitin thiolesterase 3; ubiquitin-specific-processing protease 3; UBP; Usp3
Gene Symbols USP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human USP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.