Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588817 Supplier Invitrogen™ Supplier No. PA588817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TERKKRRKGL VPGLVNLGNT CFMNSLLQGL SACPAFIRWL EEFTSQYSRD QKEPPSHQYL SLTLLHLLKA LSCQEVTDDE VLDASCLLDV LRMYRWQISS FEEQDAHELF HVITSSLEDE RDRQPRVTHL FDVHSLEQQS EITPKQITCR TRGSPHPTSN HWKSQHPFHG RLTSNMVCKH CEHQSPVRFD TFDSLSLSIP AATWGHPLTL DHCLHHFISS ESVRDVVCDN CTKIEAKGTL NGEKVEHQRT TFVKQLKLGK LPQCLCIHLQ RLSWSSHGTP LKRHEHVQFN EFLMMDIYKY HLLGHKPSQH NPKLNKNPGP TLELQDGPGA PTPV; Positive Samples: Mouse lung, Rat lung; Cellular Location: Mitochondrion outer membrane.

USP30, a member of the ubiquitin-specific protease family, is a novel mitochondrial deubiquitinating (DUB) enzyme (Nakamura and Hirose, 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP30
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene USP30
Gene Accession No. D3ZPG5, Q3UN04, Q70CQ3
Gene Alias 6330590F17Rik; AI851327; D5Ertd483e; Deubiquitinating enzyme 30; FLJ40511; MGC10702; Ubiquitin carboxyl-terminal hydrolase 30; ubiquitin specific peptidase 30; ubiquitin specific protease 30; ubiquitin thioesterase 30; ubiquitin thiolesterase 30; ubiquitin-specific protease 30; ubiquitin-specific-processing protease 30; ub-specific protease 30; USP30
Gene Symbols USP30
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100756, 304579, 84749
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.