Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106762 Supplier Invitrogen™ Supplier No. PA5106762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP30.

USP30, a member of the ubiquitin-specific protease family, is a novel mitochondrial deubiquitinating (DUB) enzyme (Nakamura and Hirose, 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP30
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP30
Gene Accession No. D3ZPG5, Q3UN04, Q70CQ3
Gene Alias 6330590F17Rik; AI851327; D5Ertd483e; Deubiquitinating enzyme 30; FLJ40511; MGC10702; Ubiquitin carboxyl-terminal hydrolase 30; ubiquitin specific peptidase 30; ubiquitin specific protease 30; ubiquitin thioesterase 30; ubiquitin thiolesterase 30; ubiquitin-specific protease 30; ubiquitin-specific-processing protease 30; ub-specific protease 30; USP30
Gene Symbols USP30
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP30(Accession Q70CQ3), corresponding to amino acid residues N34-Q84.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100756, 304579, 84749
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.