Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568355 Supplier Invitrogen™ Supplier No. PA568355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP31
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP31
Gene Accession No. Q70CQ4
Gene Alias 6330567E21Rik; Deubiquitinating enzyme 31; KIAA1203; mKIAA1203; ubiquitin carboxyl-terminal hydrolase 31; ubiquitin specific peptidase 31; ubiquitin specific protease 31; ubiquitin specific proteinase 31; ubiquitin thioesterase 31; ubiquitin thiolesterase 31; ubiquitin-specific-processing protease 31; USP31
Gene Symbols USP31
Host Species Rabbit
Immunogen Synthesized peptide derived from human USP31.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308959, 57478, 76179
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.