Learn More
Invitrogen™ USP31 Polyclonal Antibody

Description
Antibody detects endogenous levels of total USP31.
Specifications
Specifications
| Antigen | USP31 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | USP31 |
| Gene Accession No. | Q70CQ4 |
| Gene Alias | 6330567E21Rik; Deubiquitinating enzyme 31; KIAA1203; mKIAA1203; ubiquitin carboxyl-terminal hydrolase 31; ubiquitin specific peptidase 31; ubiquitin specific protease 31; ubiquitin specific proteinase 31; ubiquitin thioesterase 31; ubiquitin thiolesterase 31; ubiquitin-specific-processing protease 31; USP31 |
| Gene Symbols | USP31 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.