Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP32 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298655APC
Description
USP32 Polyclonal antibody specifically detects USP32 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
USP32 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 32, EC 3.1.2.15, EC 3.4.19.12, NY-REN-60, Renal carcinoma antigen NY-REN-60, ubiquitin carboxyl-terminal hydrolase 32, ubiquitin specific peptidase 32, ubiquitin specific protease 32, ubiquitin thioesterase 32, Ubiquitin thiolesterase 32, USP10Ubiquitin-specific-processing protease 32 | |
Produced in rabbits immunized with E. coli-derived Human USP32 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
84669 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction