Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550153
Description
The antibody detects endogenous levels of total USP32 protein.
USP32 (Ubiquitin Specific Peptidase 32) is a Protein Coding gene. Among its related pathways are Ubiquitin-Proteasome Dependent Proteolysis. Gene Ontology (GO) annotations related to this gene include calcium ion binding and thiol-dependent ubiquitin-specific protease activity. An important paralog of this gene is USP6. [GeneCards].
Specifications
USP32 | |
Polyclonal | |
Unconjugated | |
Usp32 | |
2900074J03Rik; 3.4.19.12; 6430526O11Rik; AW045245; Deubiquitinating enzyme 32; NY-REN-60; renal carcinoma antigen NY-REN-60; Ubiquitin carboxyl-terminal hydrolase 32; ubiquitin specific peptidase 32; ubiquitin specific protease 32; ubiquitin thioesterase 32; ubiquitin thiolesterase 32; ubiquitin-specific-processing protease 32; USP10; USP32 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84669 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NFA0 | |
Usp32 | |
Synthesized peptide derived from internal of human USP32 (aa 271-320). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction