Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106447 Supplier Invitrogen™ Supplier No. PA5106447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total USP33.

DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP33
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP33
Gene Accession No. Q8R5K2, Q8TEY7
Gene Alias 9830169D19Rik; AA409780; Deubiquitinating enzyme 33; hVDU1; KIAA1097; pVHL-interacting deubiquitinating enzyme 1; ubiquitin carboxyl-terminal hydrolase 33; ubiquitin specific peptidase 33; ubiquitin specific protease 33; ubiquitin thioesterase 33; ubiquitin thiolesterase 33; ubiquitin-specific-processing protease 33; Usp33; VDU1; Vhlh-interacting deubiquitinating enzyme 1; VHL-interacting deubiquitinating enzyme 1
Gene Symbols USP33
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP33(Accession Q8TEY7), corresponding to amino acid residues M292-D342.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170822, 23032, 310960
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.