Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555261 Supplier Invitrogen™ Supplier No. PA555261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHIIPFRTYV IRYLCKLSDQ ELRQSAARNM ADLMWSTVKE PLDTTLCFDK ESLDLAFKYF MSPTLTMRLA GLSQITNQLH TFNDVCNNES LVSDTETSIA KELADWLI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 21%.

Ubiquitin hydrolase that can remove conjugated ubiquitin from AXIN1 and AXIN2, thereby acting as a regulator of Wnt signaling pathway. Acts as an activator of the Wnt signaling pathway downstream of the beta-catenin destruction complex by deubiquitinating and stabilizing AXIN1 and AXIN2, leading to promote nuclear accumulation of AXIN1 and AXIN2 and positively regulate beta-catenin (CTNBB1)-mediated transcription. Recognizes and hydrolyzes the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP34
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP34
Gene Accession No. Q70CQ2
Gene Alias A530081C03Rik; deubiquitinating enzyme 34; KIAA0570; KIAA0729; Murr2; U2af1-rs1 region 2; ubiquitin carboxyl-terminal hydrolase 34; ubiquitin specific peptidase 34; ubiquitin specific protease 34; ubiquitin thioesterase 34; ubiquitin thiolesterase 34; ubiquitin-specific-processing protease 34; Usp34
Gene Symbols USP34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP34. Recombinant protein control fragment (Product #RP-89256).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.