Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP35 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128689AF700
Description
USP35 Polyclonal antibody specifically detects USP35 in Human samples. It is validated for Western BlotSpecifications
USP35 | |
Polyclonal | |
Western Blot | |
Deubiquitinating enzyme 35, EC 3.4.19.12, KIAA1372, ubiquitin carboxyl-terminal hydrolase 35, ubiquitin specific peptidase 35, ubiquitin specific protease 35, ubiquitin thioesterase 35, Ubiquitin thiolesterase 35, Ubiquitin-specific-processing protease 35, USP34 | |
The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human ubiquitin specific peptidase 35 using the numbering given in entry CAE51937.1 (GeneID 57558). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
57558 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction