Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP35 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562225
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2225 Supplier Bethyl Laboratories, Inc Supplier No. A302289A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The ubiquitin pathway involves three sequential enzymatic steps that facilitate the conjugation of ubiquitin and ubiquitin-like molecules to specific protein substrates. Through the use of a wide range of enzymes that can add or remove ubiquitin, the ubiquitin pathway controls many intracellular processes such as signal transduction, transcriptional activation and cell cycle progression. USP35 (ubiquitin specific peptidase 35) is a 1,017 amino acid protein that belongs to the peptidase C19 family. Expressed in pancreas, testis and skeletal muscle, USP35 functions to catalyze the conversion of a ubiquitin C-terminal thioester and water to free ubiquitin and a thiol, a reaction that may play a role in signaling events throughout the cell.

Specifications

Antigen USP35
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene USP35
Gene Accession No. Q9P2H5
Gene Alias Ubiquitin carboxyl-terminal hydrolase 35, Deubiquitinating enzyme 35, Ubiquitin thioesterase 35, Ubiquitin-specific-processing protease 35, KIAA1372, USP34
Gene Symbols USP35
Host Species Rabbit
Immunogen Between 875 and 925
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57558
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.