Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP36 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10040832H
Description
USP36 Polyclonal antibody specifically detects USP36 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
USP36 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Deubiquitinating enzyme 36, DUB1, EC 3.1.2.15, EC 3.4.19.12, FLJ12851, KIAA1453deubiquitinating enzyme 1, ubiquitin carboxyl-terminal hydrolase 36, ubiquitin specific peptidase 36, ubiquitin specific protease 36, ubiquitin thioesterase 36, Ubiquitin thiolesterase 36, Ubiquitin-specific-processing protease 36 | |
The immunogen recognized by this antibody maps to a region between residue 1070 and the C-terminus (residue 1121) of human Ubiquitin Specific Peptidase 36 (Deubiquinating Enzyme 1) using the numbering given in entry NP_079366.2 (GeneID 57602). | |
0.1 mL | |
Cell Biology | |
57602 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction