Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP36 Antibody (OTI7G3), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274806G
Description
USP36 Monoclonal specifically detects USP36 in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
USP36 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
USP36 | |
Recombinant protein fragment corresponding to amino acids 589-972 of human USP36 (NP_079366) produced in E.coli. | |
0.1 mL | |
57602.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI7G3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Deubiquitinating enzyme 36, DUB1, EC 3.1.2.15, EC 3.4.19.12, FLJ12851, KIAA1453deubiquitinating enzyme 1, ubiquitin carboxyl-terminal hydrolase 36, ubiquitin specific peptidase 36, ubiquitin specific protease 36, ubiquitin thioesterase 36, Ubiquitin thiolesterase 36, Ubiquitin-specific-processing protease 36 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction