Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP36 Mouse anti-Canine, Human, Clone: OTI7G3, liquid, TrueMAB™
SDP

Catalog No. 501678079
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8079 Supplier OriGene Supplier No. TA800095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 ′Lys-4′ trimethylation (H3K4). Nagase T., DNA Res. 7:143-150(2000). The MGC Project Team, Genome Res. 14:2121-2127(2004). Ota T., Nat. Genet. 36:40-45(2004).

Specifications

Antigen USP36
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USP36
Gene Accession No. Q9P275
Gene Alias DUB1
Gene Symbols USP36
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 589-972 of human USP36 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 483344, 57602
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.