Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552908 Supplier Invitrogen™ Supplier No. PA552908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSLVHSSNVK VVLNQQAYVL FYLRIPGSKK SPEGLISRTG SSSLPGRPSV IPDHSKKNIG NGIISSPLTG KRQDSGTMKK PHTTEEIGVP ISRNGSTLGL KSQNGCIPPK LPSGSPSPKL SQTPTHMPTI LDDPGKKVKK PAPP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 'Lys-4' trimethylation (H3K4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP36
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP36
Gene Accession No. Q9P275
Gene Alias 2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36
Gene Symbols USP36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP36. Recombinant protein control fragment (Product #RP-89229).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57602
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.