Learn More
Invitrogen™ USP36 Polyclonal Antibody

Description
Immunogen sequence: EDGHLKCPRSA KPQDAVVPES SSCAPSANGW CPGDRMGLSQ APPVSWNGER ESDVVQELLK YSSDKAYGRK VLTWDGKMSA VSQDAIEDSR QARTETVVDD WDEEFDRGKE KKIKKFKREK RRNFNAFQKL QTRRNFWSVT HPAK Highest antigen sequence identity to the following orthologs - mouse 70%, rat 68%.
Specifications
Specifications
| Antigen | USP36 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | USP36 |
| Gene Accession No. | Q9P275 |
| Gene Alias | 2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36 |
| Gene Symbols | USP36 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.