Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585927
Description
USP36 Polyclonal Antibody for ICC/IF, IHC (P)
May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 'Lys-4' trimethylation (H3K4).
Specifications
USP36 | |
Polyclonal | |
Unconjugated | |
USP36 | |
2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q9P275 | |
USP36 | |
Synthetic peptide within Human USP36 aa 1,074-1,123. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction