Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP36 Polyclonal Antibody, Huabio

Catalog No. 502544744
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4744 Supplier HUABIO RESEARCH INC Supplier No. ER170665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP36 Polyclonal antibody specifically detects USP36 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen USP36
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene USP36
Gene Accession No. B1AQJ2, Q9P275
Gene Alias 2700002L06Rik; deubiquitinating enzyme 36; DUB1; Kiaa1453; mKIAA1453; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific peptidase 36; ubiquitin specific protease 36; ubiquitin thioesterase 36; ubiquitin thiolesterase 36; ubiquitin-specific-processing protease 36; Usp36
Host Species Rabbit
Immunogen Recombinant protein within human PKA C alpha aa 172-338.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303700, 57602, 72344
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.