Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP37 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040696H
Description
USP37 Polyclonal antibody specifically detects USP37 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
USP37 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37 | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ubiquitin Specific Peptidase 37 using the numbering given in entry NP_065986.1 (GeneID 57695). | |
0.1 mL | |
Protein Turnover, Ubiquitin Proteasome Pathway | |
57695 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction