Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP37 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP18290125UL
Description
USP37 Polyclonal specifically detects USP37 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
USP37 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
Deubiquitinating enzyme 37, EC 3.1.2.15, EC 3.4.19.12, KIAA1594tmp_locus_50, MGC117261, ubiquitin carboxyl-terminal hydrolase 37, ubiquitin specific peptidase 37, ubiquitin specific protease 37, ubiquitin thioesterase 37, Ubiquitin thiolesterase 37, Ubiquitin-specific-processing protease 37 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
USP37 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TGITKWKEGSFEIVEKENKVSLVVHYNTGGIPRIFQLSHNIKNVVLRPSGAKQSRLMLTLQDNSFLSIDKVPSKDAEEMRLFLDAVHQNRLPAAMKPSQGSGSFGAILGSRTSQKETSR | |
25 μL | |
Protein Turnover, Ubiquitin Proteasome Pathway | |
57695 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction